Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pax6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001244198.1 | NP_001231127.1 | 11 | 1344 | ||
NM_001244200.1 | NP_001231129.1 | 12 | 1556 | |||
NM_001244201.1 | NP_001231130.1 | 10 | 1302 | |||
NM_001244202.1 | NP_001231131.1 | 10 | 1858 | |||
NM_013627.5 | NP_038655.1 | 11 | 1900 | |||
GenBank mRNA | AF443223.1 | - | 7 | 957 | ||
AF457141.1 | - | 5 | 623 | |||
AF457142.1 | - | 6 | 440 | |||
AJ292077.1 | - | 10 | 1161 | |||
AJ307468.1 | - | 10 | 1168 | |||
AK028059.1 | - | 10 | 1303 | |||
AK032616.1 | - | 11 | 1344 | |||
AK045805.1 | - | 10 | 1860 | |||
AK139054.1 | - | 888 | ||||
AY064175.1 | - | 6 | 767 | |||
BC011272.1 | - | 11 | 1588 | 4008490 | ||
BC036957.1 | - | 10 | 1298 | 4504106 | ||
BC069912.1 | - | 7 | 1335 | |||
M77842.1 | - | 6 | 887 | |||
X63963.1 | - | 11 | 1221 |