Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pax6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001244198.1 | NP_001231127.1 | 7 | 699 | ||
NM_001244200.1 | NP_001231129.1 | 8 | 911 | |||
NM_001244201.1 | NP_001231130.1 | 6 | 657 | |||
NM_001244202.1 | NP_001231131.1 | 6 | 1213 | |||
NM_013627.5 | NP_038655.1 | 7 | 1255 | |||
GenBank mRNA | AJ292077.1 | - | 6 | 516 | ||
AJ307468.1 | - | 6 | 523 | |||
AK028059.1 | - | 6 | 658 | |||
AK032616.1 | - | 7 | 700 | |||
AK045805.1 | - | 6 | 1214 | |||
AK139054.1 | - | 243 | ||||
BC011272.1 | - | 7 | 943 | 4008490 | ||
BC036957.1 | - | 6 | 653 | 4504106 | ||
M77842.1 | - | 2 | 242 | |||
X63963.1 | - | 7 | 576 |