Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcmt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008786.2 | NP_032812.2 | 1 | 488 | ||
GenBank mRNA | AK006162.1 | - | 1 | 166 | ||
AK148789.1 | - | 1 | 513 | |||
BC040750.1 | - | 1 | 81 | |||
BC049613.1 | - | 1 | 192 | |||
BC058966.1 | - | 1 | 177 | 5717963 | ||
BY099821.1 | - | 1 | 220 | |||
D38023.1 | - | 1 | 152 | |||
M60320.1 | - | 1 | 152 |