Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcsk6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291184.1 | NP_001278113.1 | 5 | 686 | ||
NM_011048.1 | NP_035178.1 | 5 | 686 | |||
GenBank mRNA | BC037450.1 | - | 3 | 291 | ||
D50060.1 | - | 5 | 609 |