Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcsk7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281934.1 | NP_001268863.1 | 8 | 1872 | ||
NM_008794.2 | NP_032820.2 | 8 | 1608 | |||
GenBank mRNA | AK148416.1 | - | 8 | 1193 | ||
AK151980.1 | - | 8 | 1195 | |||
AK165389.1 | - | 8 | 1214 | |||
BC006730.1 | - | 7 | 1109 | 3962441 | ||
BC007322.1 | - | 8 | 1471 | |||
BC056456.1 | - | 8 | 1607 | |||
U48830.1 | - | 8 | 1180 |