Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdk18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008795.2 | NP_032821.1 | 13, 14 | 1441 | ||
GenBank mRNA | AK004998.1 | - | 13, 14 | 1385 | ||
AK171569.1 | - | 13, 14 | 1222 | |||
BC119617.1 | - | 14, 15 | 1317 | 40061728 | ||
BC119618.1 | - | 8, 9 | 708 | |||
X69026.1 | - | 13, 14 | 1458 |