Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdcd11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011053.2 | NP_035183.2 | 34, 35 | 5380 | ||
GenBank mRNA | AK003899.1 | - | 3, 4 | 562 | ||
AK129080.1 | - | 34, 35 | 5372 | |||
BC010844.1 | - | 34, 35 | 5353 | |||
BC038503.1 | - | 5, 6 | 859 | |||
BC051231.1 | - | 2, 3 | 707 | |||
BC055276.1 | - | 3, 4 | 567 | |||
BC070468.1 | - | 34, 35 | 5367 | 30628812 | ||
BC094244.1 | - | 14, 15 | 2772 |