Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pde8a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008803.2 | NP_032829.1 | 1 | 221 | ||
GenBank mRNA | AF067806.1 | - | 287 | |||
AK148238.1 | - | 430 | ||||
AK149561.1 | - | 1 | 422 | |||
BC125578.1 | - | 1 | 221 | 40130093 | ||
BC132145.1 | - | 1 | 221 | 40130422 |