Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pde6g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF190928.1 | - | 3 | 219 | ||
AK080735.1 | - | 4 | 365 | |||
BC014723.1 | - | 4 | 1730 | |||
BC082288.1 | - | 4 | 322 | 6490841 | ||
CK619967.1 | - | 4 | 209 | |||
DQ092438.1 | - | 3 | 95 | |||
DQ251193.1 | - | 3 | 95 | |||
Y00746.1 | - | 4 | 340 |