Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pde6g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF190928.1 | - | 3 | 248 | ||
AK080735.1 | - | 4 | 394 | |||
BC014723.1 | - | 4 | 1759 | |||
BC082288.1 | - | 4 | 351 | 6490841 | ||
CK619967.1 | - | 4 | 238 | |||
DQ092438.1 | - | 3 | 124 | |||
DQ251193.1 | - | 3 | 124 | |||
Y00746.1 | - | 4 | 369 |