Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdpk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080773.2 | NP_001074242.1 | 3, 4 | 462 | ||
NM_001286662.1 | NP_001273591.1 | 3, 4 | 380 | |||
NM_011062.4 | NP_035192.2 | 3, 4 | 462 | |||
GenBank mRNA | AF079535.1 | - | 3, 4 | 419 | ||
AF086625.1 | - | 3, 4 | 428 | |||
AF126294.1 | - | 3, 4 | 403 | |||
AK147149.1 | - | 3, 4 | 454 | |||
AK147835.1 | - | 3, 4 | 437 | |||
AK149162.1 | - | 3, 4 | 395 | |||
AK149289.1 | - | 3, 4 | 420 | |||
AK162677.1 | - | 3, 4 | 422 | |||
AK165564.1 | - | 3, 4 | 399 | |||
AY062008.1 | - | 3, 4 | 358 | |||
BC146001.1 | - | 3, 4 | 399 | 40131304 | ||
BC146003.1 | - | 3, 4 | 399 | 40131306 | ||
CJ046953.1 | - | 3, 4 | 381 |