Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pea15a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011063.2 | NP_035193.1 | 3 | 498 | ||
GenBank mRNA | AK077421.1 | - | 3 | 427 | ||
AK089070.1 | - | 3 | 426 | |||
AK153141.1 | - | 3 | 431 | |||
AK153493.1 | - | 3 | 429 | |||
AK161635.1 | - | 3 | 414 | |||
AK161977.1 | - | 3 | 427 | |||
BC038282.1 | - | 3 | 406 | 4500957 | ||
G49234.1 | - | 3 | 420 |