Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pepd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008820.2 | NP_032846.2 | 3 | 266 | ||
GenBank mRNA | AK010581.1 | - | 3 | 247 | ||
AK145843.1 | - | 3 | 297 | |||
AK168384.1 | - | 3 | 256 | |||
BC086644.1 | - | 3 | 240 | 6807359 | ||
BC131945.1 | - | 3 | 268 | 40130222 | ||
BC131973.1 | - | 3 | 268 | 40130250 | ||
D82983.1 | - | 3 | 253 | |||
U51014.1 | - | 3 | 289 |