Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pfkfb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008824.3 | NP_032850.1 | 3 | 364 | ||
GenBank mRNA | AK132629.1 | - | 3 | 365 | ||
BC132561.1 | - | 3 | 349 | 40130838 | ||
BC132563.1 | - | 3 | 349 | 40130840 |