Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pgk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008828.3 | NP_032854.2 | 9 | 1229 | ||
GenBank mRNA | AK133877.1 | - | 9 | 1151 | ||
AK145846.1 | - | 9 | 1165 | |||
AK164440.1 | - | 9 | 1151 | |||
AK167441.1 | - | 9 | 1123 | |||
AK167459.1 | - | 9 | 1123 | |||
AK167710.1 | - | 9 | 1122 | |||
AK177443.1 | - | 3 | 364 | |||
AK188712.1 | - | 2 | 295 | |||
BC023819.1 | - | 9 | 1135 | |||
BC032167.1 | - | 9 | 1135 | |||
BC083355.1 | - | 9 | 1121 | 6828087 | ||
BC108372.1 | - | 9 | 1133 | 5060103 | ||
M15668.1 | - | 9 | 1100 |