Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fyb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK161411.1 | - | 1 | 39 |
Gene Symbol: Phb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008831.4 | NP_032857.1 | 7 | 739 | ||
GenBank mRNA | AK002714.1 | - | 6 | 685 | ||
AK010619.1 | - | 7 | 682 | |||
AK150956.1 | - | 7 | 684 | |||
AK151073.1 | - | 7 | 684 | |||
AK152933.1 | - | 7 | 686 | |||
AK168248.1 | - | 7 | 687 | |||
AK168656.1 | - | 7 | 678 | |||
BC083354.1 | - | 6 | 661 | 5715690 | ||
BC089034.1 | - | 6 | 663 | 30675988 | ||
X78682.1 | - | 6 | 667 |