Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pkm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253883.1 | NP_001240812.1 | 7 | 1213 | ||
NM_011099.3 | NP_035229.2 | 7 | 1213 | |||
GenBank mRNA | AK083076.1 | - | 7 | 1006 | ||
AK135397.1 | - | 7 | 1006 | |||
AK151724.1 | - | 7 | 1079 | |||
AK153483.1 | - | 7 | 1079 | |||
AK154695.1 | - | 7 | 1006 | |||
AK155110.1 | - | 7 | 1006 | |||
AK155655.1 | - | 7 | 1009 | |||
AK168943.1 | - | 7 | 1006 | |||
AK169872.1 | - | 7 | 947 | |||
AK170892.1 | - | 7 | 1009 | |||
AK171023.1 | - | 7 | 1006 | |||
AK171033.1 | - | 7 | 1009 | |||
AK171106.1 | - | 7 | 1010 | |||
AK190343.1 | - | 1 | 109 | |||
AK211250.1 | - | 2 | 169 | |||
AK214987.1 | - | 1 | 87 | |||
BC016619.1 | - | 7 | 973 | 3598842 | ||
BC094663.1 | - | 7 | 952 | 6829532 | ||
X97047.1 | - | 6 | 924 |