Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pla2g1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011107.1 | NP_035237.1 | 2 | 107 | ||
GenBank mRNA | AF094610.1 | - | 2 | 100 | ||
AF097637.1 | - | 2 | 88 | |||
AF162712.1 | - | 2 | 107 | |||
AF187852.1 | - | 2 | 86 | |||
AK007730.1 | - | 2 | 101 | |||
AK007797.1 | - | 2 | 106 | |||
AK008331.1 | - | 2 | 103 | |||
AK008664.1 | - | 2 | 103 | |||
AK008668.1 | - | 2 | 103 | |||
AK008841.1 | - | 2 | 101 | |||
AK008934.1 | - | 2 | 107 | |||
AK008936.1 | - | 2 | 101 | |||
AK009314.1 | - | 2 | 100 | |||
AK019009.1 | - | 2 | 101 | |||
AK028104.1 | - | 2 | 104 | |||
AK028134.1 | - | 2 | 107 | |||
BC145908.1 | - | 2 | 107 | 40131211 | ||
BC145910.1 | - | 2 | 107 | 40131213 |