Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pla2g2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001082531.1 | NP_001076000.1 | 4 | 282 | ||
GenBank mRNA | AK131857.1 | - | 4 | 303 | ||
BC045156.1 | - | 4 | 282 | 6479080 | ||
U28244.1 | - | 4 | 289 | |||
U32359.1 | - | 3 | 257 |