Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290562.1 | NP_001277491.1 | 3, 4 | 623 | ||
NM_011123.3 | NP_035253.1 | 3, 4 | 623 | |||
GenBank mRNA | AK134554.1 | - | 3, 4 | 614 | ||
AK160872.1 | - | 3, 4 | 618 | |||
BC027010.1 | - | 3, 4 | 533 | 5364736 | ||
M14674.1 | - | 2, 3 | 457 | |||
M15442.1 | - | 3, 4 | 602 | |||
M20752.1 | - | 2, 3 | 457 |