Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019755.4 | NP_062729.1 | 2 | 155 | ||
GenBank mRNA | AB031292.1 | - | 2 | 173 | ||
AK003522.1 | - | 2 | 210 | |||
AK011282.1 | - | 2 | 209 | |||
AK012816.1 | - | 2 | 250 | |||
AK154228.1 | - | 2 | 219 | |||
BC083078.1 | - | 2 | 198 | 6477560 | ||
BC119088.1 | - | 2 | 155 | 8733837 | ||
BC119090.1 | - | 2 | 155 | 8733839 |