Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pole2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011133.2 | NP_035263.1 | 3 | 258 | ||
GenBank mRNA | AF036898.1 | - | 3 | 231 | ||
AK011194.1 | - | 3 | 239 | |||
AK077592.1 | - | 3 | 259 | |||
BC052170.1 | - | 3 | 247 |