Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppib | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011149.2 | NP_035279.2 | 1, 2 | 235 | ||
GenBank mRNA | AK002357.1 | - | 1, 2 | 182 | ||
AK128977.1 | - | 1, 2 | 185 | |||
AK154073.1 | - | 1, 2 | 174 | |||
AL363449.1 | - | 1, 2 | 276 | |||
BC013061.1 | - | 1, 2 | 172 | 3483267 | ||
M60456.1 | - | 1, 2 | 154 | |||
X58990.1 | - | 1, 2 | 280 |