Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prkx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016979.1 | NP_058675.1 | 5 | 1047 | ||
GenBank mRNA | AJ238004.1 | - | 5 | 930 | ||
AK034681.1 | - | 5 | 1366 | |||
AK037141.1 | - | 5 | 1041 | |||
AK081548.1 | - | 5 | 1027 | |||
AK139510.1 | - | 5 | 1055 | |||
AK149937.1 | - | 5 | 1025 | |||
AK150588.1 | - | 5 | 1009 | |||
AK152166.1 | - | 5 | 843 | |||
AK154447.1 | - | 5 | 1027 | |||
AK169322.1 | - | 5 | 998 | |||
BC006875.1 | - | 5 | 1047 | 3596013 |