Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prodh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011172.2 | NP_035302.2 | 4 | 631 | ||
GenBank mRNA | AF120279.1 | - | 4 | 588 | ||
AK144068.1 | - | 4 | 601 | |||
BC008548.1 | - | 4 | 455 | |||
BC125327.1 | - | 4 | 434 | 40129842 | ||
U80020.1 | - | 4 | 580 |