Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pros1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011173.2 | NP_035303.1 | 6, 7 | 685 | ||
GenBank mRNA | AK145501.1 | - | 6, 7 | 685 | ||
AK163032.1 | - | 6, 7 | 713 | |||
BC128316.1 | - | 6, 7 | 647 | 40109789 | ||
L27439.1 | - | 5, 6 | 496 | |||
Z25469.1 | - | 6, 7 | 651 |