Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prpf4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013830.2 | NP_038858.2 | 8 | 2119 | ||
GenBank mRNA | AB516304.1 | - | 8 | 1971 | ||
AF033663.1 | - | 7 | 1409 | |||
AF283466.1 | - | 8 | 2117 | |||
AK083926.1 | - | 8 | 2003 | |||
BC003769.1 | - | 5 | 496 | |||
BC042521.1 | - | 9 | 1401 | |||
BC080864.1 | - | 8 | 2090 | |||
BC141272.1 | - | 8 | 2045 | 9055884 | ||
BC141273.1 | - | 8 | 2045 | 9055885 | ||
U48737.1 | - | 6 | 676 |