Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyth1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001112699.1 | NP_001106169.1 | 4 | 339 | ||
NM_001112700.1 | NP_001106170.1 | 4 | 279 | |||
NM_011180.3 | NP_035310.2 | 4 | 339 | |||
GenBank mRNA | AB013464.1 | - | 4 | 260 | ||
AF051337.1 | - | 3 | 207 | |||
AK078502.1 | - | 250 | ||||
AK158216.1 | - | 4 | 279 | |||
AK172296.1 | - | 4 | 284 | |||
AK220302.1 | - | 4 | 261 | |||
BC057974.1 | - | 4 | 223 | 5060913 | ||
BY204859.1 | - | 4 | 279 | |||
CJ131185.1 | - | 4 | 339 |