Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psmb4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008945.3 | NP_032971.2 | 5, 6 | 737 | ||
GenBank mRNA | AK077448.1 | - | 5, 6 | 714 | ||
AK137044.1 | - | 5, 6 | 713 | |||
AK161348.1 | - | 5, 6 | 713 | |||
AK168265.1 | - | 5, 6 | 713 | |||
AK168340.1 | - | 5, 6 | 713 | |||
BC008241.1 | - | 5, 6 | 692 | 2654421 | ||
U65636.1 | - | 5, 6 | 692 |