Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psmc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008948.2 | NP_032974.2 | 8 | 1073 | ||
GenBank mRNA | AK002542.1 | - | 8 | 794 | ||
AK083043.1 | - | 1035 | ||||
AK151370.1 | - | 8 | 793 | |||
AK152018.1 | - | 8 | 812 | |||
AK167002.1 | - | 8 | 1021 | |||
AK167061.1 | - | 8 | 1018 | |||
AK168263.1 | - | 7 | 742 | |||
AK168616.1 | - | 8 | 793 | |||
BC005783.1 | - | 8 | 962 | 3603053 | ||
D49686.1 | - | 8 | 1017 |