Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pipox | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008952.2 | NP_032978.2 | 2 | 294 | ||
GenBank mRNA | AK008555.2 | - | 2 | 255 | ||
AK135009.1 | - | 1 | 1627 | |||
AK149488.1 | - | 2 | 243 | |||
BC013525.1 | - | 2 | 208 | 4237443 | ||
BY708439.1 | - | 2 | 255 | |||
U94700.1 | - | 2 | 206 |