Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pstpip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011193.2 | NP_035323.2 | 10 | 1126 | ||
GenBank mRNA | AK037936.1 | - | 9 | 1928 | ||
AK088150.1 | - | 9 | 1928 | |||
U87814.1 | - | 10 | 966 |