Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pstpip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011193.2 | NP_035323.2 | 8, 9 | 999 | ||
GenBank mRNA | BC096761.1 | - | 8, 9 | 779 | 30040955 | |
U87814.1 | - | 8, 9 | 839 |