Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bcas3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166642.1 | NP_001160114.1 | 16 | 1396 | ||
NM_001166643.1 | NP_001160115.1 | 15 | 1310 | |||
NM_138681.4 | NP_619622.3 | 16 | 1396 | |||
GenBank mRNA | AF313800.1 | - | 15 | 1290 | ||
AK032423.1 | - | 16 | 1347 | |||
AK034117.1 | - | 16 | 1376 | |||
AK160127.1 | - | 15 | 1289 | |||
BC028339.1 | - | 15 | 1284 | |||
BC038173.1 | - | 14 | 1253 | |||
BC050233.1 | - | 15 | 1289 | |||
BC060690.1 | - | 15 | 1273 | |||
BC080775.1 | - | 14 | 1257 | |||
BC118965.2 | - | 15 | 1310 | 40097104 | ||
BC118966.1 | - | 15 | 1292 | 40097105 |