Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wfdc15b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001045554.1 | NP_001039019.1 | 3 | 260 | ||
NM_138685.2 | NP_619626.1 | 3 | 292 | |||
GenBank mRNA | AF276974.1 | - | 3 | 289 | ||
AK078998.1 | - | 3 | 240 | |||
AK085392.1 | - | 3 | 228 | |||
BB625483.1 | - | 3 | 237 | |||
BC061041.1 | - | 3 | 237 | 6740598 | ||
CB954017.1 | - | 3 | 263 |