Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008973.2 | NP_032999.1 | 2 | 1489 | ||
GenBank mRNA | AK011346.1 | - | 2 | 346 | ||
AK011699.1 | - | 2 | 272 | |||
AK017768.1 | - | 2 | 308 | |||
AK133835.1 | - | 2 | 289 | |||
AK161847.1 | - | 2 | 239 | |||
AK161964.1 | - | 2 | 179 | |||
BC002064.1 | - | 2 | 58 | 3488877 | ||
BC048363.1 | - | 2 | 63 | |||
BC061695.1 | - | 2 | 289 | 6313227 | ||
D90225.1 | - | 2 | 233 |