Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptprc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111316.2 | NP_001104786.2 | 10 | 1110 | ||
NM_001268286.1 | NP_001255215.1 | 7 | 719 | |||
NM_011210.4 | NP_035340.3 | 8 | 791 | |||
GenBank mRNA | AK054056.1 | - | 8 | 792 | ||
AK154893.1 | - | 7 | 714 | |||
BC131655.1 | - | 8 | 731 | |||
L36091.1 | - | 3 | 317 | |||
M14342.1 | - | 8 | 753 | |||
M14343.1 | - | 4 | 1284 | |||
M92933.1 | - | 10 | 1110 |