Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rassf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138956.3 | NP_620406.1 | 3, 4 | 583 | ||
GenBank mRNA | AF332864.1 | - | 3, 4 | 594 | ||
AK155495.1 | - | 3, 4 | 610 | |||
AK167627.1 | - | 3, 4 | 416 | |||
BC011511.1 | - | 3, 4 | 583 | 3495833 | ||
BC055023.1 | - | 3, 4 | 638 | 5250629 | ||
BC071204.1 | - | 3, 4 | 591 | 2648929 |