Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rapgef6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252494.1 | NP_001239423.1 | 1, 2 | 273 | ||
NM_001252496.1 | NP_001239425.1 | 1, 2 | 273 | |||
NM_175258.4 | NP_780467.2 | 1, 2 | 273 | |||
GenBank mRNA | AK045540.1 | - | 1, 2 | 215 | ||
AK049228.1 | - | 1, 2 | 229 | |||
AK167055.1 | - | 1, 2 | 274 | |||
BC141182.1 | - | 1, 2 | 172 | 9055794 | ||
BC145503.1 | - | 1, 2 | 172 |