Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rapgef6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252494.1 | NP_001239423.1 | 24, 25 | 3827 | ||
NM_001252496.1 | NP_001239425.1 | 25, 26 | 3851 | |||
NM_175258.4 | NP_780467.2 | 23, 24 | 3812 | |||
GenBank mRNA | AK143331.1 | - | 11, 12 | 2199 | ||
AK220459.1 | - | 13, 14 | 2239 | |||
BC019702.1 | - | 2, 3 | 389 | |||
BC025553.1 | - | 6, 7 | 1306 | |||
BC059847.1 | - | 6, 7 | 1232 | |||
BC092525.1 | - | 6, 7 | 1379 | |||
BC141182.1 | - | 24, 25 | 3726 | 9055794 | ||
BC145503.1 | - | 25, 26 | 3751 |