Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyb5d2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024926.3 | NP_001020097.2 | 2 | 431 | ||
GenBank mRNA | AK034043.1 | - | 2 | 129 | ||
BC086682.1 | - | 2 | 397 | 30666422 | ||
BC117721.1 | - | 1 | 13 |