Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pxn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011223.3 | NP_035353.1 | 6 | 909 | ||
NM_133915.3 | NP_598676.2 | 6 | 909 | |||
GenBank mRNA | AF293882.1 | - | 6 | 834 | ||
AF293883.1 | - | 5 | 729 | |||
AK084243.1 | - | 4 | 574 | |||
AK148060.1 | - | 6 | 895 | |||
AK149933.1 | - | 6 | 874 | |||
AK167299.1 | - | 6 | 848 | |||
AK171226.1 | - | 6 | 897 | |||
AK171436.1 | - | 6 | 874 | |||
BC003298.1 | - | 2 | 192 | |||
BC025493.1 | - | 6 | 867 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK089534.1 | - | 2788 | Chr. - - [-] on Build |