Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009000.3 | NP_033026.1 | 5 | 692 | ||
GenBank mRNA | AB232619.1 | - | 5 | 366 | ||
AK007602.1 | - | 5 | 654 | |||
AK160309.1 | - | 4 | 735 | |||
BC019950.1 | - | 5 | 646 | 4011304 | ||
BC028872.1 | - | 5 | 598 | 4220908 | ||
BC054466.1 | - | 5 | 657 | 6530844 | ||
BC055894.1 | - | 5 | 655 | 3964529 | ||
BC055914.1 | - | 3 | 928 | |||
Z22819.1 | - | 5 | 366 |
Gene Symbol: Prelid1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC059038.1 | - | 309 |