Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam65b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080381.1 | NP_001073850.1 | 3, 4 | 561 | ||
NM_001286100.1 | NP_001273029.1 | 3, 4 | 429 | |||
NM_001286101.1 | NP_001273030.1 | 4, 5 | 654 | |||
NM_029679.2 | NP_083955.1 | 3, 4 | 561 | |||
NM_178658.5 | NP_848773.3 | 3, 4 | 414 | |||
GenBank mRNA | AK031928.1 | - | 3, 4 | 514 | ||
AK036452.1 | - | 4, 5 | 634 | |||
AK039271.1 | - | 3, 4 | 526 | |||
AK040887.1 | - | 3, 4 | 394 | |||
AK041665.1 | - | 3, 4 | 415 | |||
AK122269.1 | - | 3, 4 | 493 | |||
AK134538.1 | - | 3, 4 | 492 | |||
AK163880.1 | - | 3, 4 | 497 | |||
BB638362.1 | - | 3, 4 | 410 | |||
BC059179.1 | - | 3, 4 | 429 |