Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rad1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289447.1 | NP_001276376.1 | 3, 4 | 416 | ||
NM_001289448.1 | NP_001276377.1 | 3, 4 | 628 | |||
NM_011232.3 | NP_035362.2 | 3, 4 | 628 | |||
GenBank mRNA | AF038841.1 | - | 3, 4 | 539 | ||
AF058394.1 | - | 2, 3 | 315 | |||
AF073523.1 | - | 3, 4 | 555 | |||
AF074718.1 | - | 2, 3 | 325 | |||
AF076842.1 | - | 2, 3 | 326 | |||
AF084514.1 | - | 3, 4 | 487 | |||
AJ004976.1 | - | 2, 3 | 337 | |||
AK146533.1 | - | 3, 4 | 598 | |||
AK148098.1 | - | 3, 4 | 587 | |||
AK150466.1 | - | 3, 4 | 537 | |||
AK164168.1 | - | 2, 3 | 786 | |||
AK168588.1 | - | 3, 4 | 578 | |||
BC048693.1 | - | 2, 3 | 392 | 6508142 |