Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rad21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009009.4 | NP_033035.3 | 4, 5 | 610 | ||
GenBank mRNA | AF332085.1 | - | 3, 4 | 365 | ||
AF332086.1 | - | 3, 4 | 365 | |||
AK004746.1 | - | 4, 5 | 413 | |||
AK128984.1 | - | 4, 5 | 499 | |||
AK139492.1 | - | 3, 4 | 263 | |||
AK145591.1 | - | 4, 5 | 577 | |||
AK163992.1 | - | 471 | ||||
AK168718.1 | - | 4, 5 | 598 | |||
AK168896.1 | - | 4, 5 | 559 | |||
BC043032.1 | - | 3, 4 | 379 | 5696540 | ||
BC129916.1 | - | 3, 4 | 409 | 40111820 | ||
BC129917.1 | - | 3, 4 | 410 | 40111821 | ||
D37790.1 | - | 3, 4 | 297 | |||
X98293.1 | - | 3, 4 | 365 |