Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rag2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009020.3 | NP_033046.1 | 3 | 1230 | ||
GenBank mRNA | AK031014.1 | - | 3 | 1240 | ||
AK040375.1 | - | 3 | 1231 | |||
AK089075.1 | - | 3 | 1262 | |||
BC120548.1 | - | 3 | 1187 | 40129471 | ||
BC125473.1 | - | 3 | 1187 | 40129988 | ||
BC144856.1 | - | 2 | 1124 | 9053404 | ||
M64796.1 | - | 3 | 1174 |
Gene Symbol: B230118H07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | DQ112091.1 | - | 291 |