Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rag2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009020.3 | NP_033046.1 | 3 | 1446 | ||
GenBank mRNA | AK031014.1 | - | 3 | 1456 | ||
AK040375.1 | - | 3 | 1447 | |||
AK089075.1 | - | 3 | 1478 | |||
BC120548.1 | - | 3 | 1403 | 40129471 | ||
BC125473.1 | - | 3 | 1403 | 40129988 | ||
BC144856.1 | - | 2 | 1340 | 9053404 | ||
M64796.1 | - | 3 | 1390 |