Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rasgrp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011242.2 | NP_035372.2 | 14, 15 | 1724 | ||
GenBank mRNA | AK160563.1 | - | 4, 5 | 455 | ||
Y12339.1 | - | 6, 7 | 543 |