Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rasgrf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039655.1 | NP_001034744.1 | 1 | 292 | ||
NM_011245.2 | NP_035375.1 | 1 | 292 | |||
GenBank mRNA | AF169826.1 | - | 1 | 146 | ||
AK038936.1 | - | 1 | 156 | |||
AK079638.1 | - | 1 | 168 | |||
AK140227.1 | - | 1 | 159 | |||
AK140380.1 | - | 1 | 159 | |||
AK162908.1 | - | 1 | 159 | |||
BC042449.1 | - | 1 | 159 | |||
BC138682.1 | - | 1 | 128 | 8861703 | ||
BC138683.1 | - | 1 | 128 | 8861704 | ||
L20899.1 | - | 1 | 294 |